ContactUsLinkComponent
Human Retinoic Acid Receptor beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quant itative determination of human Retinoic Acid Receptor beta in tissue homogenates, cell lysates or other biological fl 1 * 48 Tests
Catalog # ANTIA7423-48
Supplier:  Antibodies.com
New Product
Human Retinoic Acid Receptor beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quant itative determination of human Retinoic Acid Receptor beta in tissue homogenates, cell lysates or other biological fl 1 * 48 Tests
Catalog # ANTIA7423-48
Supplier:  Antibodies.com