ContactUsLinkComponent
Mouse Retinoid X Receptor alpha/RXRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Retinoid X Receptor alpha/RXRA in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78752-96
Supplier:  Antibodies.com
New Product
Mouse Retinoid X Receptor alpha/RXRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Retinoid X Receptor alpha/RXRA in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA78752-96
Supplier:  Antibodies.com