ContactUsLinkComponent
Mouse Adenosine A2b Receptor/ADORA2B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Adenosine A2b Receptor/ADORA2B in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA7432-48
Supplier:  Antibodies.com
New Product
Mouse Adenosine A2b Receptor/ADORA2B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Adenosine A2b Receptor/ADORA2B in tissue homogenates, cell lysates or other biologi 1 * 48 Tests
Catalog # ANTIA7432-48
Supplier:  Antibodies.com