ContactUsLinkComponent
Mouse VDAC1/Porin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse VDAC1/Porin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA82393-96
Lieferant:  Antibodies.com
New Product
Mouse VDAC1/Porin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse VDAC1/Porin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 96 Tests
Catalog # ANTIA82393-96
Lieferant:  Antibodies.com