ContactUsLinkComponent
Human alpha 1,2 Mannosidase IA/MAN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of human alpha 1,2 Mannosidase IA/MAN1A1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6868-96
Supplier:  Antibodies.com
New Product
Human alpha 1,2 Mannosidase IA/MAN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of human alpha 1,2 Mannosidase IA/MAN1A1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6868-96
Supplier:  Antibodies.com