ContactUsLinkComponent
Rat Cytochrome P450 1A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of rat Cytochrome P450 1A2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6439-96
Supplier:  Antibodies.com
New Product
Rat Cytochrome P450 1A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of rat Cytochrome P450 1A2 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6439-96
Supplier:  Antibodies.com