ContactUsLinkComponent
Rat Epithelial Sodium Channel alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Epithelial Sodium Channel alpha in serum, plasma, tissue homogenates or other biologic 1 * 96 Tests
  ANTIA7040-96
New Product
Rat Epithelial Sodium Channel alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Epithelial Sodium Channel alpha in serum, plasma, tissue homogenates or other biologic 1 * 96 Tests
  ANTIA7040-96