Human Adenosine A2b Receptor/ADORA2B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Adenosine A2b Receptor/ADORA2B in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA86934-96
Supplier: Antibodies.com