ContactUsLinkComponent
Mouse Advanced Glycation End Products/AGE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of mouse Advanced Glycation End Products/AGE in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA77648-96
Supplier:  Antibodies.com
New Product
Mouse Advanced Glycation End Products/AGE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of mouse Advanced Glycation End Products/AGE in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA77648-96
Supplier:  Antibodies.com