ContactUsLinkComponent
Human Interferon gamma Receptor beta/AF-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Interferon gamma Receptor beta/AF-1 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA73845-96
Supplier:  Antibodies.com
New Product
Human Interferon gamma Receptor beta/AF-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Interferon gamma Receptor beta/AF-1 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA73845-96
Supplier:  Antibodies.com