ContactUsLinkComponent
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitr o quantitative determination of human Bone Marrow Stromal Cell Antigen 1 in serum, plasma or other biological fluids. 1 * 48 Tests
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit
Catalog # ANTIA6223-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit
Catalog # ANTIA6223-48
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Frequently Bought Together