Rat ZO1 Tight Junction Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat ZO1 Tight Junction Protein in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8046-96
Supplier: Antibodies.com