ContactUsLinkComponent
11-Nor-9beta-hydroxyhexahydrocannabinol (11-Nor-9beta-HHC) solution 1 mg/mL (Acetonitrile (Degas)), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-166-1ML
Supplier:  Merck
New Product
11-Nor-9beta-hydroxyhexahydrocannabinol (11-Nor-9beta-HHC) solution 1 mg/mL (Acetonitrile (Degas)), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-166-1ML
Supplier:  Merck
New Product