ContactUsLinkComponent
24,25-Dihydroxyvitamin D3 solution 50 µg/mL (Degassed Ethanol with 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-160-1ML
Supplier:  Merck
New Product
24,25-Dihydroxyvitamin D3 solution 50 µg/mL (Degassed Ethanol with 0.1% BHT), certified reference material, ampule of 1 mL, Cerilliant 1 * 1 mL
Catalog # H-160-1ML
Supplier:  Merck
New Product