ContactUsLinkComponent
Be 1000mg/l in diluted HNO3 for ICP/MS CRM, ISO 17034 and ISO 17025 Conservation 36 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSDM207.2NP.L1
Supplier:  CPACHEM
New Product
Be 1000mg/l in diluted HNO3 for ICP/MS CRM, ISO 17034 and ISO 17025 Conservation 36 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSDM207.2NP.L1
Supplier:  CPACHEM