Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2-Bromophenyl)ethanol 98%

2-(2-Bromophenyl)ethanol 98%

Bottle
Catalog # APOSOR480688-500G
Irritant
New Product
Supplier:   Apollo Scientific
2-(2-Bromophenyl)ethanol 98%
Catalog # APOSOR480688-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  1074-16-4
2-(2-Bromophenyl)ethanol 98%

Bottle

CAS Number:  1074-16-4
Price
885,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:1074-16-4
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00093566
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:97 °C/0.675mmHg
  • Density:1.483 g/cm³ (25 °C)
  • Flash Pt:>113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications