Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Chloro-6-nitrophenol

2-Chloro-6-nitrophenol

Bottle
Catalog # APOSOR400451-500G
Irritant
New Product
Supplier:   Apollo Scientific
2-Chloro-6-nitrophenol
Catalog # APOSOR400451-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  603-86-1
2-Chloro-6-nitrophenol

Bottle

CAS Number:  603-86-1
Price
681,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:603-86-1
  • Formula:C₆H₄ClNO₃
  • MDL Number:MFCD00024233
  • Molecular Weight:173.56 g/mol
  • Melting Pt:67…70 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.