Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-Formyl-5-methoxypyridine

Bottle
Catalog # APOSOR313046-5G
Irritant
New Product
Supplier:   Apollo Scientific
2-Formyl-5-methoxypyridine
Catalog # APOSOR313046-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  22187-96-8
2-Formyl-5-methoxypyridine

Bottle

CAS Number:  22187-96-8
Price
197,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde
  • CAS Number:22187-96-8
  • Formula:C₇H₇NO₂
  • MDL Number:MFCD08234867
  • Molecular Weight:137.1360 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.