Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Pyrocatechol 99%

Bottle
Catalog # APOSOR923744-50G
New Product
Supplier:   Apollo Scientific
Pyrocatechol 99%
Catalog # APOSOR923744-50G
Supplier:  Apollo Scientific
New Product
CAS Number:  120-80-9
Pyrocatechol 99%

Bottle

CAS Number:  120-80-9
Price
47,80 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Boiling Pt:245 °C
  • Melting Pt:103…106 °C
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications