ContactUsLinkComponent
LN² Capacity (L): 35l Outer Diameter (mm): 448 Overall Height (mm): 668 Neck Opening (mm): 119 Weight Empty (kg): 17,3 Weight Full (kg): 45,5 Evaporation Rate: 0,27 Number of Canisters: 6 Static Holding Time: 130 1,2 & 2ml Vial (6/cane): 1260 1/2cc S 1 * 1 ST
  TAYW0020003
New Product
LN² Capacity (L): 35l Outer Diameter (mm): 448 Overall Height (mm): 668 Neck Opening (mm): 119 Weight Empty (kg): 17,3 Weight Full (kg): 45,5 Evaporation Rate: 0,27 Number of Canisters: 6 Static Holding Time: 130 1,2 & 2ml Vial (6/cane): 1260 1/2cc S 1 * 1 ST
  TAYW0020003
New Product