ContactUsLinkComponent
35 Liter, w/10 Canisters CCF-Series Model CCF35H CCF35V LN2 Capacity (L) 35 Outer Diameter (in/mm) 18.8 (448) Overall Height (in/mm) 26.3 (668) Neck Opening (in/mm) 4.7 (119) Canister Dims. (in/mm) 2.64x11 (67x279) Weight Full (lbs/kg) 101 (45.9) Wei 1 * 1 ST
Catalog # TAYW0020001
Supplier:  TAYLOR WHARTON
New Product
35 Liter, w/10 Canisters CCF-Series Model CCF35H CCF35V LN2 Capacity (L) 35 Outer Diameter (in/mm) 18.8 (448) Overall Height (in/mm) 26.3 (668) Neck Opening (in/mm) 4.7 (119) Canister Dims. (in/mm) 2.64x11 (67x279) Weight Full (lbs/kg) 101 (45.9) Wei 1 * 1 ST
Catalog # TAYW0020001
Supplier:  TAYLOR WHARTON
New Product