Auftragserfassung
Germany
AuftragsverfolgungKontakt
Valinomycin

Valinomycin

Bottle
Catalog # APOSBIV0893-500MG
Toxic
Supplier:   Apollo Scientific
Valinomycin
Catalog # APOSBIV0893-500MG
Supplier:  Apollo Scientific
CAS Number:  2001-95-8
Valinomycin

Bottle

CAS Number:  2001-95-8
Price
817,00 €
Log in for availability
1 * 500 mg
Product Details & Documents

Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions

  • CAS Number:2001-95-8
  • Formula:C₅₄H₉₀O₁₈N₆
  • MDL Number:MFCD00005114
  • Molecular Weight:1111.34 g/mol
  • Melting Pt:187…190 °C
  • Storage Temperature:Kühlschrank
  • EINECS:217-896-6
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.