Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrocatechol Violet

Pyrocatechol Violet

Bottle
Catalog # APOSBIP0720-100G
Irritant
Supplier:   Apollo Scientific
Pyrocatechol Violet
Catalog # APOSBIP0720-100G
Supplier:  Apollo Scientific
CAS Number:  115-41-3
Pyrocatechol Violet

Bottle

CAS Number:  115-41-3
Price
553,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Pyrocatechol Violet
  • CAS Number:115-41-3
  • Formula:C₁₉H₁₄O₇S
  • MDL Number:MFCD00005868
  • Molecular Weight:386,37526 g/mol
  • EINECS:204-088-3
Specifications

Specifications

  • Pack type:
    Bottle

Specifications