Order Entry
Germany
Orders LinkContactUsLinkComponent
L-Methionine ≥99%

L-Methionine ≥99%

Bottle
Catalog # APOSBIM0715-2.5KG
New Product
Supplier:   Apollo Scientific
L-Methionine ≥99%
Catalog # APOSBIM0715-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  63-68-3
L-Methionine ≥99%

Bottle

CAS Number:  63-68-3
Price
485,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Ph.Eur, Pyrogen free

Synonyms:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS Number:63-68-3
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00063097
  • Molecular Weight:149.21 g/mol
  • Melting Pt:284 °C
  • Storage Temperature:Ambient
  • EINECS:200-562-9

Dokumente

Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.