Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol

Bottle
Catalog # APOSBI2889-100G
Irritant
Supplier:   Apollo Scientific
D-(-)-Sorbitol
Catalog # APOSBI2889-100G
Supplier:  Apollo Scientific
CAS Number:  50-70-4
D-(-)-Sorbitol

Bottle

CAS Number:  50-70-4
Price
43,50 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient

 

Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.