Ectoine 98%
Bottle
Catalog # APOSOR97119-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
- CAS Number:96702-03-3
- Formula:C₆H₁₀N₂O₂
- MDL Number:MFCD03419286
- Molecular Weight:142,157 g/mol
- EINECS:431-910-1
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



