Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2-Chlorophenyl)propionic acid 97%

2-(2-Chlorophenyl)propionic acid 97%

Bottle
Catalog # APOSOR962317-5G
New Product
Supplier:   Apollo Scientific
2-(2-Chlorophenyl)propionic acid 97%
Catalog # APOSOR962317-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  2184-85-2
2-(2-Chlorophenyl)propionic acid 97%

Bottle

CAS Number:  2184-85-2
Price
401,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-(2-Chlorophenyl)propanoic acid
  • CAS Number:2184-85-2
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD06658208
  • Molecular Weight:184.62 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.