ContactUsLinkComponent
pH 10.00 at 20 C coloured Borax/Sodium Hydroxide CRM, ISO 17034 and ISO 17025 Conservation 24 months Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # ACSDPH010.L5
Supplier:  CPACHEM
New Product
pH 10.00 at 20 C coloured Borax/Sodium Hydroxide CRM, ISO 17034 and ISO 17025 Conservation 24 months Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # ACSDPH010.L5
Supplier:  CPACHEM