Order Entry
Germany
Orders LinkContactUsLinkComponent
Basic Red 9

Basic Red 9

Bottle
Catalog # APOSOR1012314-500G
Supplier:   Apollo Scientific
Basic Red 9
Catalog # APOSOR1012314-500G
Supplier:  Apollo Scientific
CAS Number:  569-61-9
Basic Red 9

Bottle

CAS Number:  569-61-9
Price
71,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
  • CAS Number:569-61-9
  • Formula:C₁₉H₁₇N₃·ClH
  • MDL Number:MFCD00001657
  • Molecular Weight:323,82 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.