Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-(4-Chlorophenoxy)benzaldehyde 97%

Bottle
Catalog # APOSOR91108-1G
Supplier:   Apollo Scientific
2-(4-Chlorophenoxy)benzaldehyde 97%
Catalog # APOSOR91108-1G
Supplier:  Apollo Scientific
CAS Number:  111826-11-0
2-(4-Chlorophenoxy)benzaldehyde 97%

Bottle

CAS Number:  111826-11-0
Price
84,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:111826-11-0
  • Formula:C₁₃H₉ClO₂
  • MDL Number:MFCD01568734
  • Molecular Weight:232,66 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications