Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(4-Chlorophenoxy)benzaldehyde 97%

2-(4-Chlorophenoxy)benzaldehyde 97%

Bottle
Catalog # APOSOR91108-100G
Supplier:   Apollo Scientific
2-(4-Chlorophenoxy)benzaldehyde 97%
Catalog # APOSOR91108-100G
Supplier:  Apollo Scientific
CAS Number:  111826-11-0
2-(4-Chlorophenoxy)benzaldehyde 97%

Bottle

CAS Number:  111826-11-0
Price
4 020,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:111826-11-0
  • Formula:C₁₃H₉ClO₂
  • MDL Number:MFCD01568734
  • Molecular Weight:232,66 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.