Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OBJECTIVE 60X XPLAN APO OIL NA 1.42 WD 0.15MM 1 * 1 items
ECHLJMAG8601
:
DISCOVER ECHO
New Product
OBJECTIVE 60X XPLAN APO OIL NA 1.42 WD 0.15MM 1 * 1 items
ECHLJMAG8601
:
DISCOVER ECHO
New Product