ContactUsLinkComponent
Vertical outer 560h x 138w x 142d with 5 apertures of 111mm supplied complete with fibre rod and locking rod. 6 racks required to fill IC Biomedical LS6000 1 * 1 ST
Catalog # TAYW002070
Supplier:  TAYLOR WHARTON
New Product
Vertical outer 560h x 138w x 142d with 5 apertures of 111mm supplied complete with fibre rod and locking rod. 6 racks required to fill IC Biomedical LS6000 1 * 1 ST
Catalog # TAYW002070
Supplier:  TAYLOR WHARTON
New Product