Aluminum Sample Pans, 80 µL (pkg. of 100): TGA aluminum sample pans for use in the Q5000 IR and Discovery TGA 55, 550, 5500 over the temperature range of ambient to 600°C. Note: Stainless Steel Bail (P/N 957364.901) is required to use the aluminum sa 1 * 100 ST
Catalog # MSPP957363901
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together