Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1X PBS with 0.05% Tween 20, 1% BSA, 1000mL, Sterile 1 * 1.000 mL
Catalog # TEKOP0234
Supplier:
TEKNOVA
Expand image
1X PBS with 0.05% Tween 20, 1% BSA, 1000mL, Sterile 1 * 1.000 mL
Catalog # TEKOP0234
Supplier:
TEKNOVA