Precellys lysing kit - hard tissue homohgenizing ck14 (1.4mm zirconium oxide beads) - 2 ml 1 * 50 KIT
Catalog # BERTP000912LYSK1A0
Supplier: Bertin Instruments
New Product
Precellys lysing kit - hard tissue homohgenizing ck14 (1.4mm zirconium oxide beads) - 2 ml 1 * 50 KIT
Catalog # BERTP000912LYSK1A0
Supplier: Bertin Instruments
New Product
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Starch (from wheat), powder, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Accessories for VWR Bead Mill Homogenisers
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific