TopTes Guard-101 Gas Detector, 4 Gas Detectors for H2S, CO, LEL and O2, with Vibration, Light and Sound Alarm, 14 Hours Battery Life , Explosion-Proof, for Work and Home - Blue 1 * 1 ST
Catalog # MSPPB0D7C5T65W
Supplier: MARKET SOURCE PART PROCESS