ContactUsLinkComponent
Ashing furnace lv15/11/b510 tmax1100 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELV155K11NLTY
Supplier:  NABERTHERM
New Product
Ashing furnace lv15/11/b510 tmax1100 with flap door and over-temperature limiter and thermocouple hole in the furnace door 1 * 1 items
Catalog # NABELV155K11NLTY
Supplier:  NABERTHERM