b.safe Laboratory Flasks
500 ml, GL 45
Catalog # BOHLR105-79
Supplier: Bohlender
Jump to:
Product Details & Documents
Laboratory flask made of borosilicate glass.
Includes screw cap and spout made of PP
Specifications
- Thread:GL 45
- Capacity:500 ml