Column-Refill up to 150mm x 4,6mm ID -NA - Refill of PH columns like HITA855-4506 #please send two old columns per refill to #Frank Gutjahr Chromatographie Hauptstrasse 48 72336 Balingen Germany #add reference HPLCS00005S-NA / VWR 1 * 1 items
Catalog # HPLCS00005S-NA
Supplier: COLLECTION PREFIX HPLC
New Product