Buffered NaCl Peptone Solution pH 7,0
Buffered NaCl peptone solution, 7,0, 90 ml, 6
Catalog # LIOF402730
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Buffered NaCl Peptone Solution pH 7,0 is used for dissolving, suspending and diluting test samples. Its composition conforms to recommendations in the Harmonised Pharmacopoeias for the microbiological examination of non-sterile products.
Zertifizierung: This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics.
Bestellinformationen: For professional use only.
Specifications
Specifications
- Pk:6
- Description:Buffered NaCl peptone solution
- Filling:90 ml
- pH value:7,0
- Cat. No.:LIOF402730
- Storage temperature:10...25 °C



