Order Entry
Germany
Orders LinkContactUsLinkComponent
Human PBMC

Human PBMC

Human PBMC, healthy donor, discovery, 40M cells (40×10⁶ cells/vial), Cryopreserved, Single donor, 1 Vial
Catalog # PRTV12193102040
Supplier:   PROVITRO
Human PBMC
Catalog # PRTV12193102040
Supplier:  PROVITRO
CAS Number:  
Human PBMC
Human PBMC

Human PBMC, healthy donor, discovery, 40M cells (40×10⁶ cells/vial), Cryopreserved, Single donor, 1 Vial

CAS Number:  
Price
1 090,00 €
Log in for availability
1 * 1 Vial
Product Details & Documents

Human peripheral blood mononuclear cells (PBMCs) are utilised as source material for isolating lymphocytes (T cells, B cells and NK cells) and monocytes in research and across fields such as immunology, immuno-oncology, and cell and gene therapies.
All PBMCs are isolated through Ficoll density gradient separation from freshly obtained leukapheresis ACD-A packs and directly cryopreserved.

  • PBMC-standard includes basic donor information and serological tests
  • Additional optional information and tests are available for an extra fee
  • Cells are cryopreserved in CryoStor® CS10 media
  • Purity is >90% as determined by flow cytometry

Our offerings encompass distinct options tailored to meet diverse research requirements. These options include PBMCs sourced from our existing inventory, which are further categorised into three groups, each offering unique features and application possibilities.

We also provide PBMCs from prospective collections, ensuring flexibility and suitability for various research needs. Storage: For prolonged cell preservation or resuscitation, it is recommended to prepare the cells for extended storage in the vapor phase of liquid nitrogen. Storage of cells in the liquid phase of nitrogen is strongly discouraged to prevent potential cross-contamination risks when removing from storage. Long-term storage under −175 °C and for short-term storage, about one or two weeks at −80 °C is permitted, but it is recommended to minimise this duration to ensure optimal stability. Thawed samples should be used immediately and not stored.

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1 Vial
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human PBMC, healthy donor, discovery, 40M cells (40×10⁶ cells/vial)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.