Orders LinkContactUsLinkComponent
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Achromat N.A. 0,40, 1,5 mm
Catalog # ECHLRMAG02022
Supplier:   DISCOVER ECHO
Objectives for Rebel Microscopes
Catalog # ECHLRMAG02022
Supplier:  DISCOVER ECHO
Objectives for Rebel Microscopes
Objectives for Rebel Microscopes
Achromat N.A. 0,40, 1,5 mm
Price
281,00 €
Log in for availability
1 * 1 ST
Specifications
  • Magnification:
    20×
  • Description:
    Achromat N.A. 0,40
  • Seq:
    14
  • Working distance:
    1,5 mm