Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
18 components: 100mg/l each of As Ba Cd Co Cr Cu Mn Ni Pb Sb Se V Zn Al Ag Fe Mo Tl in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM113221L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
18 components: 100mg/l each of As Ba Cd Co Cr Cu Mn Ni Pb Sb Se V Zn Al Ag Fe Mo Tl in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM113221L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product