Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution no.2 (yellow) for the examination of the degree of colouration of liquids in accordance with the Chinese pharmacopoeia, Glass bottle, 100 mL
Catalog # REGECPRCSY201
Supplier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalog # REGECPRCSY201
Supplier:  REAGECON
CAS Number:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution no.2 (yellow) for the examination of the degree of colouration of liquids in accordance with the Chinese pharmacopoeia, Glass bottle, 100 mL

CAS Number:  
Price
189,00 €
Log in for availability
1 * 100 mL
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Colour reference solution no.2 (yellow) for the examination of the degree of colouration of liquids in accordance with the Chinese pharmacopoeia

Specifications