Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Al 10mg/l As 10mg/l Cu 10mg/l Mo 10mg/l Se 10mg/l Sr 10mg/l Ti 10mg/l Zn 100mg/l in HNO3 3% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM112555L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
Al 10mg/l As 10mg/l Cu 10mg/l Mo 10mg/l Se 10mg/l Sr 10mg/l Ti 10mg/l Zn 100mg/l in HNO3 3% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # CMPLRM112555L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product