1.6 mL 96-Well Deep Well Plate, U-Bottom, Square Well, Non-Sterile, PP 5/pk, 50/cs Pyrogens: Non-Pyrogenic RNase/DNase: DNase/RNase free BSE/TSE: These products are deemed animal free 1 * 50 ST
Catalog # MSPP502002
Supplier: MARKET SOURCE PART PROCESS