PIG® Essentials Universal Socks
PIG® essentials universal socks, 94,6 L/pack, Grey, 80×3660 mm
Catalog # NEWPMS1210
Supplier: NEW PIG
Jump to:
Product Details & Documents
Polypropylene-filled socks and booms to absorb oils, coolants, solvents and water.
- 100% polypropylene-filled socks and booms absorb oils, coolants, solvents and water
- Ideal for surrounding spilled fluids or wrapping around leaky machines
- Easy and convenient to put into place; contain spills to keep fluids from spreading into larger areas and becoming slippery hazards
- Flexible to conform around machine bases and offer a snug fit with no seepage
- Grey colour signifies Universal sorbency for easy visual identification
- Contains 100% post-industrial recycled poly blend filler
The product consists of a polypropylene absorbent. Contained within a casing made of: Polypropylene (CAS 9003-07-0). Also containing impregnated: Surfactant. Pigment. May contain: Mixture Adhesive; Polypropylene film; Polyester yarn.
Specifications
- Colour:Grey
- Description:PIG® essentials universal socks
- Seq:02
- Absorbency:94,6 L/pack
- Ø×L:80×3660 mm



