Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 96 HE with reflector module FL EC P and C (E), consisting of the filters EX BP 390/40, BS FT 420, EM BP 450/40 installed in reflector module with reflection prevention through inclined mount for emission filter
Catalog # ZEIS4890969100400
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4890969100400
Supplier:  ZEISS
Filter sets
Filter set 96 HE with reflector module FL EC P and C (E), consisting of the filters EX BP 390/40, BS FT 420, EM BP 450/40 installed in reflector module with reflection prevention through inclined mount for emission filter
Price
2 008,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 96 HE with reflector module FL EC P and C (E), consisting of the filters EX BP 390/40, BS FT 420, EM BP 450/40 installed in reflector module with reflection prevention through inclined mount for emission filter