HEK293/Human APP GFP Stable Cell Line 1 * 2 Vial
Catalog # ACRBCHEK-ATP081-2
Supplier: ACRO Biosystems
HEK293/Human APP GFP Stable Cell Line 1 * 2 Vial
Catalog # ACRBCHEK-ATP081-2
Supplier: ACRO Biosystems
Frequently Bought Together
[EN]MARCING DYE BLUE 1 * 59 mL
Supplier: BRADLEY
Sodium hydroxide 1 mol/l, EMPROVE® EXPERT, SAFC®
Supplier: Merck
[EN]IGG1 ISOTYPE CONTROL N297Q 1 * 5 mg
Supplier: MARKET SOURCE PART PROCESS
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
NON DENATURED ETHANOL 70% 1 * 1 L
Supplier: PROLABO CMP
Colorimetric rapid tests, phosphate, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Ferrous Ammonium Sulphate 0.1M Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
CeO2-2 1 * 1 kg
Supplier: MARKET SOURCE PART PROCESS
Hyaluronate Mono Cydye, MW 5 kDa 1 * 50 mg
Supplier: MARKET SOURCE PART PROCESS
HEK293/Human LDL R Stable Cell Line 1 * 2 Vial
Supplier: ACRO Biosystems